missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiomethoxide, 90+%, technical

Product Code. 17281336 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17281336 100 g 100g
17251336 1 g 1g
17261336 5 g 5g
17271336 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17281336 Supplier Thermo Scientific Acros Supplier No. 469891000
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5188-07-8 | CH3NaS | 70.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5188-07-8
Molecular Formula CH3NaS
Molecular Weight (g/mol) 70.08
InChI Key RMBAVIFYHOYIFM-UHFFFAOYSA-M
IUPAC Name sodium methylsulfanide
SMILES [Na+].C[S-]

Specifications

Assay Percent Range 90+%
Linear Formula CH3SNa
Quantity 100 g
Solubility Information Solubility aq. soln.: freely soluble.
Formula Weight 70.09
Grade Technical
Chemical Name or Material Sodium thiomethoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.