missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiomethoxide, 95%, pure

Product Code. 10361044 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10361044 1 g Glass bottle 1g
10043710 5 g Glass bottle 5g
10217223 25 g Glass bottle 25g
11955741 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10361044 Supplier Thermo Scientific Acros Supplier No. 305710010
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5188-07-8 | CH3NaS | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5188-07-8
Molecular Formula CH3NaS
Molecular Weight (g/mol) 70.09
MDL Number MFCD00174316
InChI Key RMBAVIFYHOYIFM-UHFFFAOYSA-M
PubChem CID 4378561
SMILES C[S-].[Na+]

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 94 to 106% (Iodimetry)
Packaging Glass bottle
Linear Formula CH3SNa
Quantity 1 g
Synonym sodium thiomethoxide, sodium methanethiolate, sodium thiomethylate, methyl mercaptan sodium salt, methanethiol sodium salt, sodiumthiomethoxide, methanethiol, sodium salt, sodiummethanethiolate, sodium methyl mercaptide, methylsulfanyl sodium
Solubility Information Solubility in water: freely soluble.
IUPAC Name sodium;methanethiolate
Formula Weight 70.09
Percent Purity 95%
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium thiomethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.