missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, 99%, for analysis, anhydrous

Product Code. 15219656 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15219656 5 kg 5kg
15213406 250 g 250g
15271906 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15219656 Supplier Thermo Scientific Acros Supplier No. 450620050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7772-98-7 ∣ Na2O3S2 ∣ 158.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Melting Point 48.0°C
pH 6.0 to 8.4 (5% aq. soln.)
Physical Form Crystalline Powder
Quantity 5 kg
Assay Percent Range 98.5% min. (Iodimetry)
Linear Formula Na2S2O3
Merck Index 15,8821
Solubility Information Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium sulfanidesulfonate
Formula Weight 158.1
Percent Purity 99%
Grade Analytical
Loss on Drying 1% max. (105°C)
Packaging Plastic Bottle
Chemical Name or Material Sodium thiosulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.