missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, anhydrous, 98+%

Product Code. 11438267
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438267 2500 g 2500g
11428267 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438267 Supplier Thermo Scientific Alfa Aesar Supplier No. A17629.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7772-98-7 ∣ Na2O3S2 ∣ 158.10 g/mol

Sodium thiosulfate is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used to prepare S-alkylthiosulfonates. Furthermore, it is used in the tanning of leather and in photographic processing. It is used as an antidote to cyanide poisoning and acts as a safe disposal medium in chemistry laboratories when working with bromine, iodine and other strong oxidizers. It is an important component in hand warmers and heating pads.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Sodium thiosulfate is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used to prepare S-alkylthiosulfonates. Furthermore, it is used in the tanning of leather and in photographic processing. It is used as an antidote to cyanide poisoning and acts as a safe disposal medium in chemistry laboratories when working with bromine, iodine and other strong oxidizers. It is an important component in hand warmers and heating pads.

Solubility

Soluble in water. Insoluble in alcohol.

Notes

Hygroscopic. Incompatible with strong acids, strong oxidizing agents, iodine and mercury.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Physical Form Crystalline Powder and/or Crystals
Quantity 2500 g
Linear Formula Na2S2O3
Merck Index 14,8694
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 158.1
Percent Purity 99%
Grade Ungraded
Odor Odorless
Sensitivity Hygroscopic
Density 1.667
Chemical Name or Material Sodium thiosulfate, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.