missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, 99.5+%, ACS reagent

Product Code. 10732964 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10732964 5 kg Plastic bottle 5kg
10697411 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10732964 Supplier Thermo Scientific Acros Supplier No. 424460051
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-17-7
MDL Number MFCD00149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Melting Point 48.5°C
Color White
pH 6.0 to 8.4 (5% soln. at 25°C)
Physical Form Crystals
Quantity 5 kg
Assay Percent Range 99.5 to 101% (Iodimetry)
Molecular Formula Na2O3S2·5H2O
Linear Formula Na2S2O3·5H2O
Merck Index 15, 8821
Solubility Information Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in turpentine
Molecular Weight (g/mol) 248.18
Formula Weight 248.18
Percent Purity ≥99.5%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 1.72
Decomposition Information >45°C
Density 1.7200g/mL
Chemical Name or Material Sodium thiosulfate pentahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.