missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium thiosulfate pentahydrate, 99+%, for biochemistry

Product Code. 12665127 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12665127 250 g 250g
12655127 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12665127 Supplier Thermo Scientific Chemicals Supplier No. 447942500
  • ONLINE EXCLUSIVE PRICE
    59.75 €
    56.00 € / 250g
    Save 3.75 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10102-17-7 ∣ Na2O3S2·5H2O ∣ 248.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-17-7
Molecular Formula Na2O3S2·5H2O
Molecular Weight (g/mol) 248.18
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Melting Point 48.5°C
Physical Form Powder, Crystals and/or Chunks
Quantity 250 g
Packaging Plastic bottle
Specific Gravity 1.72
Assay Percent Range 99.0 to 101% (Iodimetry)
Density 1.7200g/mL
Linear Formula Na2S2O3·5H2O
Merck Index 15,8821
Solubility Information (1 M in water) Clear colorless
Formula Weight 248.18
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Sodium thiosulfate pentahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.