missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Pentahydrate, puriss., Solstice
BIOPHARMA

Product Code. 15661350
Change view
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
4 x 5 kg
6 x 2.5 kg
Unit Size:
2.5kg
500g
5kg
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15661350 5 kg 5kg
15651350 500 g 500g
15631350 2.5 kg 2.5kg
15671350 6 x 2.5 kg Pack of 6
15641350 4 x 5 kg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15661350 Supplier Honeywell-Fluka Supplier No. 134795KG
  • ONLINE EXCLUSIVE PRICE
    188.00 €
    169.27 € / 5kg
    Save 18.73 €
    9% Off
 Restricted Item
Estimated Shipment: 14-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, European Pharmacopoeia, USP, 99-101%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
MDL Number MFCD00149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Physical Form Crystalline Solid
Quantity 5 kg
Linear Formula Na2S2O3 · 5H2O
UN Number NONH for all modes of transport
Formula Weight 248.18
Percent Purity 99 to 101%
Grade puriss.
Chemical Name or Material Sodium Thiosulfate Pentahydrate, Meets analytical specification of Ph. Eur.BP, USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.