missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Solution, 0.0394M, Honeywell Fluka™

Product Code. 15684850 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Unit Size:
1 L
10 L
5 L
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15684850 10 L 10 L
15604860 1 L 1 L
15624860 5 L 5 L
15614860 5 L 5 L
15634860 6 x 1 L Pack of 6
15694850 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15684850 Supplier Honeywell Fluka Supplier No. 3522410L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volumetric, 0.0394 M Na2S2O3 (0.0394N), for the determination of iodine adsorption number according to ASTM D 1510

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Quantity 10 L
Linear Formula Na2S2O3
UN Number NONH for all modes of transport
IUPAC Name disodium sulfanidesulfonate
Formula Weight 158.11
Density 1.01g/mL at 20°C
Chemical Name or Material Sodium thiosulfate solution, volumetric, 0.0394 M Na2S2O3 (0.0394N), for the determination of iodine adsorption number according to ASTM D 1510
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.