missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulphate pentahydrate, BAKER ANALYZED™, J.T. Baker™

Product Code. 10132550 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10132550 1 L 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10132550 Supplier JT Baker Supplier No. 0319.1000
  • ONLINE EXCLUSIVE PRICE
    83.07 €
    71.15 € / 1kg
    Save 11.92 €
    14% Off
 Restricted Item
Estimated Shipment: 08-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets ACS Specifications

Specifications

CAS 10102-17-7
pH 6.0 to 8.4 (5% Solution at 25°C)
Quantity 1 L
Assay Percent Range 99.5 to 101.0%
Molecular Formula H10Na2O8S2
MDL Number MFCD00149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium pentahydrate sulfanidesulfonate
Molecular Weight (g/mol) 248.17
Packaging Plastic bottle for solids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.