missing translation for 'onlineSavingsMsg'
Learn More

Sodium tin(IV) oxide trihydrate, 96%

Product Code. 11488527
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11488527

Brand: Thermo Scientific Alfa Aesar A18611.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12209-98-2 | H6Na2O6Sn | 266.732 g/mol

Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tin(IV) oxide trihydrate is used in the preparation of boscage-like stannic oxide by facile one-step hydrothermal route.

Solubility
Soluble in water. Insoluble in ethanol and acetone.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12209-98-2
Molecular Formula H6Na2O6Sn
Molecular Weight (g/mol) 266.732
MDL Number MFCD00150189
InChI Key SFXJSNATBHJIDS-UHFFFAOYSA-N
Synonym sodium stannate trihydrate, unii-nj7c1v83kg, nj7c1v83kg, sodium stannate inci, sodium stannate vandf, disodium stannate, trihydrate, sodium stannate iv trihydrate mi, 2na.so3n.3h2o, sodiumstannatetrihydrate, sodium tin iv oxide trihydrate
PubChem CID 21977542
IUPAC Name disodium;dioxido(oxo)tin;trihydrate
SMILES O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]

Specifications

Quantity 100 g
Merck Index 14,8677
Solubility Information Soluble in water. Insoluble in ethanol and acetone.
Formula Weight 266.75 (212.67 Anhydrous)
Percent Purity 98%
Odor Odorless
Chemical Name or Material Sodium tin(IV) oxide trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.