missing translation for 'onlineSavingsMsg'
Learn More

Sodium Trifluoroacetate, Honeywell™

Product Code. 15631580 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15631580 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15631580 Supplier Honeywell-Fluka Supplier No. 1784010G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2923-18-4
Quantity 10 g
Molecular Formula C2F3NaO2
Linear Formula CF3COONa
MDL Number MFCD00013217
UN Number NONH for all modes of transport
Beilstein 3597949
Synonym sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
InChI Key UYCAUPASBSROMS-UHFFFAOYSA-M
SMILES [Na+].[O-]C(=O)C(F)(F)F
IUPAC Name sodium trifluoroacetate
Molecular Weight (g/mol) 136.01
PubChem CID 517019
Formula Weight 136.01g/mol
Percent Purity ≥99%
Density 1.49g/mL
Chemical Name or Material Sodium Trifluoroacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.