missing translation for 'onlineSavingsMsg'
Learn More

Sodium tripolyphosphate, for analysis

Product Code. 10229243 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10229243

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-29-4 | Na5O10P3 | 367.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-29-4
Molecular Formula Na5O10P3
Molecular Weight (g/mol) 367.85
MDL Number MFCD00003514
InChI Key HWGNBUXHKFFFIH-UHFFFAOYSA-I
Synonym sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n
PubChem CID 24455
IUPAC Name pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate
SMILES [O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 622.0°C
Color White
pH 9.8 to 10.2 (1% soln.)
Physical Form Crystalline Powder
Quantity 500 g
Linear Formula Na5P3O10
Merck Index 15, 8824
Solubility Information Solubility in water: 145g/L (20°C)
Formula Weight 367.85
Percent Purity ≥98.0%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium tripolyphosphate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.