missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tungstate Dihydrate, 98-102%, Extra Pure, SLR, Fisher Chemical

Product Code. 10579390 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10579390 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10579390 Supplier Fisher Chemical Supplier No. S/7320/48
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Specifications

CAS 10213-10-2
Color White
pH 8 to 9
Physical Form Solid
Quantity 100 g
Molecular Formula H4Na2O6W
MDL Number 149190
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
Molecular Weight (g/mol) 329.846
PubChem CID 150191
ChEBI CHEBI:63939
Formula Weight 329.85g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Sodium tungstate dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.