missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tungstate Dihydrate, ACS Reagent, Solstice

Product Code. 15612200 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15612200 500 g 500g
15622200 5 g 5g
15602200 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15612200 Supplier Honeywell-Fluka Supplier No. 223336500G
  • ONLINE EXCLUSIVE PRICE
    606.00 €
    287.37 € / 500g
    Save 318.63 €
    52% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10213-10-2
Molecular Formula H4Na2O6W
Molecular Weight (g/mol) 329.846
MDL Number MFCD00149190
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
PubChem CID 150191
ChEBI CHEBI:63939
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Linear Formula Na2WO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 329.85g/mol
Percent Purity ≥99%
Grade Not Available
Chemical Name or Material Sodium Tungstate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.