missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tungstate Dihydrate, Honeywell™

Product Code. 15648180 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Packaging:
Poly bottle
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15648180 50 g Poly bottle 50g
15628180 100 g Poly bottle 100g
15638180 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15648180 Supplier Honeywell Research Chemicals Supplier No. T262950G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10213-10-2
Molecular Formula H4Na2O6W
Molecular Weight (g/mol) 329.846
MDL Number MFCD00149190
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
PubChem CID 150191
ChEBI CHEBI:63939
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 698°C
Color Colorless
pH 9.0 to 11.5 (at 100.00g/L 20°C)
Quantity 50 g
Linear Formula Na2WO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 329.85g/mol
Odor Odorless
Packaging Poly bottle
Appearance Crystalline
Density 4.18g/cu.cm
Chemical Name or Material Sodium tungstate dihydrate, Suitable for preparation of protein-free filtrates according to Folin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.