missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tungstate Dihydrate, purum, Honeywell Fluka™

Product Code. 15611420 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
6 x 500 g
Unit Size:
100g
500g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15611420 500 g 500g
15601420 100 g 100g
15621420 6 x 500 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15611420 Supplier Honeywell-Fluka Supplier No. 14304500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purum,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10213-10-2
Molecular Formula H4Na2O6W
Molecular Weight (g/mol) 329.846
MDL Number MFCD00149190
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
PubChem CID 150191
ChEBI CHEBI:63939
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 500 g
Linear Formula Na2WO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 329.85g/mol
Percent Purity ≥99%
Grade purum
Chemical Name or Material Sodium Tungstate Dihydrate
compliance-icons
Product Identifier
  • Sodium tungstate dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium tungstate-2-hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.