missing translation for 'onlineSavingsMsg'
Learn More

Sodium tungsten oxide dihydrate, ACS, 99.0-101.0%

Product Code. 11387948
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387948 500 g 500g
11397948 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387948 Supplier Thermo Scientific Alfa Aesar Supplier No. 036489.36
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10213-10-2 | H4Na2O6W | 329.846 g/mol

In organic chemistry, sodium tungstate is used as catalyst for epoxidation of alkenes and oxidation of alcohols into aldehydes or ketones. Used in examining the serum protein, serum protein-free filtrate. Used as alkaloid precipitating agent. Used in manufacture of metal tungsten, tungstate and tungsten oxide. Waterproof and fireproof fabric.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10213-10-2
Molecular Formula H4Na2O6W
Molecular Weight (g/mol) 329.846
MDL Number MFCD00149190
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
PubChem CID 150191
ChEBI CHEBI:63939
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 100°C (in -2H2O)
Color White
Physical Form Powder
Quantity 500 g
Merck Index 14,8698
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 329.86 (293.83 Anhydrous)
Percent Purity 99.0 to 101.0%
Grade ACS
Odor Odorless
Density 3.250 g/mL
Chemical Name or Material Sodium tungsten oxide dihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.