missing translation for 'onlineSavingsMsg'
Learn More

Sodium Valproate, TRC

Product Code. 30138668
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138668 10 g 10g
30135032 25 g 25g
30041368 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138668 Supplier Toronto Research Chemicals Supplier No. TRCS67450010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8 H15 O2 . Na
Molecular Weight (g/mol) 166.19
Synonym Sodium Valproate, 2-Propylpentanoic acid sodium salt (1:1), 2-Propylvaleric acid sodium salt, Acediprol, Convulex, Depacon, Depakene, Depakene R, Depakene syrup 8, Depakin, Depakine chronosphere, Dipropylacetate sodium, Epilim, KW 6066N, Orfiril, Selenica R, Sodium α,α-dipropylacetate, Sodium 2-propylpentanoate, Sodium 2-propylvalerate, Sodium bispropylacetate, Sodium dipropylacetate, Sodium valproate, Valerin, Valproate sodium
IUPAC Name sodium;2-propylpentanoate
SMILES [Na+].CCCC(CCC)C(=O)[O-]

Specifica

Quantity 10 g
InChI Formula InChI=1S/C8H16O2.Na/c1-3-5-7(6-4-2)8(9)10;/h7H,3-6H2,1-2H3,(H,9,10);/q;+1/p-1
Formula Weight 166.097
Percent Purity >95%
Chemical Name or Material Sodium Valproate
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.