missing translation for 'onlineSavingsMsg'
Learn More

Sodium vinylsulfonate, tech. ca 25% aq. soln.

Product Code. 11308256
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308256 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11308256 Supplier Thermo Scientific Alfa Aesar Supplier No. L15356.AE
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3039-83-6 | C2H3NaO3S | 130.09 g/mol

Sodium vinylsulfonate is a useful reagent (monomer) for the formation of poly(anionic) polymers and copolymers. It is employed as a basic brightener and leveling agent in nickel baths. It is also used as intermediate for organic synthesis, surfactant, pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium vinylsulfonate is a useful reagent (monomer) for the formation of poly(anionic) polymers and copolymers. It is employed as a basic brightener and leveling agent in nickel baths. It is also used as intermediate for organic synthesis, surfactant, pharmaceutical industry.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3039-83-6
Molecular Formula C2H3NaO3S
Molecular Weight (g/mol) 130.09
MDL Number MFCD00007520
InChI Key BWYYYTVSBPRQCN-UHFFFAOYSA-M
Synonym sodium ethenesulfonate, sodium vinylsulfonate, ethenesulfonic acid, sodium salt, sodium ethylenesulfonate, sodium vinyl sulfonate, sodium ethylenesulphonate, sodium apolate, unii-f7k3l38z7b, sodium ethenesulfonate in h2o, lyapolate sodium usan
PubChem CID 3270262
IUPAC Name sodium;ethenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C=C

Specifications

Density 1.18
Refractive Index 1.377
Quantity 100 mL
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 130.1
Grade Technical
Concentration or Composition (by Analyte or Components) ≈25% aq. soln.
Chemical Name or Material Sodium vinylsulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.