missing translation for 'onlineSavingsMsg'
Learn More

Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Black

Product Code. 17742088
Change view
Click to view available options
Length (Metric):
91 cm
150 cm
1830 cm
Width (Metric):
60 cm
91 cm
122 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
17742088 150 cm 91 cm Each
17702088 91 cm 60 cm Each
17782068 1830 cm 60 cm Each
17722078 1830 cm 91 cm Each
17762078 1830 cm 122 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. Vea la política de devoluciones
Código de producto. 17742088 Proveedor Notrax N.º de proveedor 411S0335BL
  • ONLINE EXCLUSIVE PRICE
    78.28 €
    63.55 € / Pack of 1
    Save 14.73 €
    18% Off
Estimated Shipment: 25-11-2026
Añadir a la cesta
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.

  • Pebble pattern provides non-directional traction and is easy to clean
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards

Especificaciones

Color Black
Length (English) 59 in.
Length (Metric) 150 cm
Material Microcellular Vinyl
Type Industrial and Commercial Matting
Width (Metric) 91 cm
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.