missing translation for 'onlineSavingsMsg'
Learn More

Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Black/Yellow

Product Code. 17762088
Change view
Click to view available options
Dimensions (L x W):
91 x 60 cm
150 x 91 cm
1830 x 60 cm
1830 x 91 cm
1830 x 122 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17762088 150 x 91 cm Each
17722088 91 x 60 cm Each
17702078 1830 x 60 cm Each
17742078 1830 x 91 cm Each
17782078 1830 x 122 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17762088 Supplier Notrax Supplier No. 411S0335BY
  • ONLINE EXCLUSIVE PRICE
    79.80 €
    69.08 € / Pack of 1
    Save 10.72 €
    13% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.

  • Pebble pattern provides non-directional traction and is easy to clean
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards

Specifications

Dimensions (L x W) 150 x 91 cm
Color Black/Yellow
Length (English) 59 in.
Length (Metric) 150 cm
Material Microcellular Vinyl
Type Industrial and Commercial Matting
Width (Metric) 91 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.