missing translation for 'onlineSavingsMsg'
Learn More

Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Gray

Product Code. 17732088
Change view
Click to view available options
Length (Metric):
91 cm
150 cm
1830 cm
Width (Metric):
60 cm
91 cm
122 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) unitSize
17732088 91 cm 60 cm Each
17772088 150 cm 91 cm Each
17712078 1830 cm 60 cm Each
17752078 1830 cm 91 cm Each
17792078 1830 cm 122 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17732088 Supplier Notrax Supplier No. 411S0323GY
  • ONLINE EXCLUSIVE PRICE
    31.50 €
    27.67 € / Pack of 1
    Save 3.83 €
    12% Off
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.

  • Pebble pattern provides non-directional traction and is easy to clean
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards

Specifications

Color Gray
Length (English) 36 in.
Length (Metric) 91 cm
Material Microcellular Vinyl
Type Industrial and Commercial Matting
Width (Metric) 60 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.