missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir impurity J, MedChemExpress

Product Code. 16492469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Packungsgröße:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
16492469 1 mg 1mg
16421258 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16492469 Lieferant Medchem Express Lieferanten-Nr. HYI09751mg
 Restricted Item
Estimated Shipment: 01-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MedChemExpress Sofosbuvir impurity J, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

Sofosbuvir impurity J, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

Spezifikation

Chemical Name or Material Sofosbuvir impurity J
CAS 1334513-10-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H30FN4O8P
Quantity 1 mg
SMILES O[C@H]1[C@](F)(C)[C@H](N2C=CC(N)=NC2=O)O[C@@H]1CO[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C)C)=O)=O
Molecular Weight (g/mol) 528.47
Formula Weight 528.47
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Mehr anzeigen Weniger anzeigen

Research purposes only

Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.