missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir impurity M, MedChemExpress

Product Code. 16462469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462469 1 mg 1mg
16401258 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462469 Supplier Medchem Express Supplier No. HYI0735
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sofosbuvir impurity M, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

Sofosbuvir impurity M, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.

Specifications

Chemical Name or Material Sofosbuvir impurity M
CAS 2095551-10-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H30N3O10P
Quantity 1 mg
SMILES O=[P@@](N[C@@H](C)C(OC(C)C)=O)(OC[C@@H]1[C@@H](O)[C@@](O)(C)[C@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3
Molecular Weight (g/mol) 527.46
Formula Weight 527.46
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.