missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir, MedChemExpress

Product Code. 16496616
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496616 100 mg 100mg
16492407 50 mg 50mg
16456288 10 mg 10mg
16411756 500 mg 500mg
16408877 5 mg 5mg
16401469 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16496616 Supplier Medchem Express Supplier No. HY15005
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sofosbuvir (GS-7977) is an HCV RNA replication inhibitor with an EC50 of 92 nM.

Sofosbuvir (GS-7977) is an HCV RNA replication inhibitor with an EC50 of 92 nM.

Specifications

Chemical Name or Material Sofosbuvir
CAS 1190307-88-0
Color Brown
Purity Grade Notes Research
Molecular Formula C22H29FN3O9P
Quantity 100 mg
Synonym GS-7977 PSI-7977
Solubility Information DMSO : 100 mg/mL (188.88 mM; Need ultrasonic) ∣H2O : 25 mg/mL (47.22 mM; ultrasonic and warming and heat to 50°C)
SMILES O=C1N([C@H]2[C@]([C@H](O)[C@@H](CO[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C)C)=O)=O)O2)(C)F)C=CC(N1)=O
Molecular Weight (g/mol) 529.45
Formula Weight 529.45
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.