Learn More
Thermo Scientific™ SoftFit-L™ Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers
Description
Thermo Scientific™ART SoftFit-L™ empty racks, when used in conjunction with SoftFit-L reload towers and inserts, offer an environmentally friendly packaging option with less plastic waste than traditional racks. Compatible with ART SoftFit-L reload inserts and reload towers.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Model | SoftFit L Rack |
| Quantity | 10 Racks/Pk., 5 Pks./Cs. |
| Product Type | Pipet Tip Rack |
| For Use With (Equipment) | 20 μL Pipet Tip, 200 μL Pipet Tip, 300 μL Pipet Tip |
| Product Line | ART SoftFit |