Learn More
Thermo Scientific™ SOLA™ SPE Cartridges
Product Code. 16654804 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
SOLA HRP SPE Cartridge
SOLA HRP SPE Cartridge 30 mg/3 mL
SOLA SAX SPE Cartridge
SOLA SAX SPE Cartridge 30 mg/3 mL
SOLA SCX SPE Cartridge
SOLA SCX SPE Cartridge 30 mg/3 mL
SOLA WAX SPE Cartridge
SOLA WAX SPE Cartridge 30 mg/3 mL
SOLA WCX SPE Cartridge
SOLA WCX SPE Cartridge 30 mg/3 mL
Sorbent:
HRP (Reversed Phase Polymeric)
Mixed Mode Strong Anion Exchanger, Polymeric
Mixed Mode Strong Cation Exchanger, Polymeric
Mixed Mode Weak Anion Exchanger, Polymeric
Mixed Mode Weak Cation Exchanger, Polymeric
Unit Size:
Pack of 100
Pack of 50
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Sorbent Unit Size
16654804 SOLA SCX SPE Cartridge 30 mg/3 mL Mixed Mode Strong Cation Exchanger, Polymeric Pack of 50
11879163 SOLA HRP SPE Cartridge HRP (Reversed Phase Polymeric) Pack of 100
16644804 SOLA HRP SPE Cartridge 30 mg/3 mL HRP (Reversed Phase Polymeric) Pack of 50
11869163 SOLA SCX SPE Cartridge Mixed Mode Strong Cation Exchanger, Polymeric Pack of 100
11889163 SOLA SAX SPE Cartridge Mixed Mode Strong Anion Exchanger, Polymeric Pack of 100
16664804 SOLA SAX SPE Cartridge 30 mg/3 mL Mixed Mode Strong Anion Exchanger, Polymeric Pack of 50
15169653 SOLA WCX SPE Cartridge Mixed Mode Weak Cation Exchanger, Polymeric Pack of 100
16674804 SOLA WCX SPE Cartridge 30 mg/3 mL Mixed Mode Weak Cation Exchanger, Polymeric Pack of 50
15189653 SOLA WAX SPE Cartridge Mixed Mode Weak Anion Exchanger, Polymeric Pack of 100
16684804 SOLA WAX SPE Cartridge 30 mg/3 mL Mixed Mode Weak Anion Exchanger, Polymeric Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 16654804 Supplier Thermo Scientific™ Supplier No. 60409002
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Grow your laboratory productivity. Thermo Scientific™ SOLA™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
More about SOLA SPE cartridges
SOLA products are available in a 2mg/1mL, 10mg/1mL and 10mg/3mL cartridge formats. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of SOLA cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
SOLA cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What SOLA Cartridges Mean to Your Analysis
  • Prevent sample failures
  • Prevent blocked wells or cartridges
  • Reduced reanalysis
TRUSTED_SUSTAINABILITY
Specifications
Description SOLA SCX SPE Cartridge 30 mg/3 mL
Bed Weight 30 mg
For Use With (Application) Bioanalytical Applications - Strong Ion-Exchange Retention of Weak Basic Compounds. Complementary Reversed Phase Retention of Neutral Compounds.
Product Line SOLA
Sorbent Mixed Mode Strong Cation Exchanger, Polymeric
Type SPE Cartridge
Volume (Metric) 3 mL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.