Learn More
Thermo Scientific™ SOLA™ SPE Plates
Product Code. 16697845 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
10 mg/2 mL
30 mg/2 mL
Sorbent:
HRP (Reversed Phase Polymeric)
Mixed Mode Strong Anion Exchanger, Polymeric
Mixed Mode Strong Cation Exchanger, Polymeric
Mixed Mode Weak Anion Exchanger, Polymeric
Mixed Mode Weak Cation Exchanger, Polymeric
Unit Size:
1 pack
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Sorbent Unit Size
16697845 30 mg/2 mL HRP (Reversed Phase Polymeric) 1 pack
11899163 10 mg/2 mL HRP (Reversed Phase Polymeric) Each
11809173 10 mg/2 mL Mixed Mode Strong Cation Exchanger, Polymeric Each
15547495 10 mg/2 mL Mixed Mode Strong Cation Exchanger, Polymeric Each
16607855 30 mg/2 mL Mixed Mode Strong Cation Exchanger, Polymeric 1 pack
15550163 10 mg/2 mL Mixed Mode Strong Anion Exchanger, Polymeric Each
11819173 10 mg/2 mL Mixed Mode Strong Anion Exchanger, Polymeric Each
16617855 30 mg/2 mL Mixed Mode Strong Anion Exchanger, Polymeric 1 pack
13493038 10 mg/2 mL Mixed Mode Weak Cation Exchanger, Polymeric Each
16627855 30 mg/2 mL Mixed Mode Weak Cation Exchanger, Polymeric 1 pack
15179653 10 mg/2 mL Mixed Mode Weak Anion Exchanger, Polymeric Each
16637855 30 mg/2 mL Mixed Mode Weak Anion Exchanger, Polymeric 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 16697845 Supplier Thermo Scientific™ Supplier No. 60509001
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Innovative Thermo Scientific™ SOLA™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
  • Higher levels of reproducibility
  • Lower elution volumes
  • High extract cleanliness
More about SOLA SPE 96 well plates
SOLA SPE Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You’ll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
Phase Types
  • Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX)
  • Strong Anion Exchanger (SAX), Weak Cation Exchanger (WCX)
  • Weak Anion Exchanger (WAX)
Bed Weights
  • 10 mg/2 mL 96-well plate format
  • 30 mg/2 ml 96-well plate format
SOLA products are also available in cartridge format
TRUSTED_SUSTAINABILITY
Specifications
Description 30 mg/2 mL
Bed Weight 30 mg
For Use With (Application) Bioanalytical Applications - Hydrophobic Retention of Neutral Compounds with Complementary Retention of Polar Analytes. An All-Purpose Phase.
Product Line SOLA
Sorbent HRP (Reversed Phase Polymeric)
Type 96-well SPE Plate
Volume (Metric) 2 mL
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.