Learn More
Thermo Scientific™ Solid Door Option for Large Chamber
Product Code. 16471125 Shop All Thermo Scientific Products
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
16471125 Each
1 options
Product Code. 16471125 Supplier Thermo Scientific™ Supplier No. 1900665
  • ONLINE EXCLUSIVE PRICE
    1960.00 €
    1870.00 € / Pack of 1
    Save 90.00 €
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Protection for your light sensitive samples
Solid outer door for Thermo Scientific™ chamber - replaces standard outer glass door to protect samples against light. Recommended for light sensitive applications. Factory-installed option, please order in conjunction with environmental chamber. F
  • Replaces standard outer glass door to protect samples against light
  • Recommended for light sensitive applications
  • Factory-installed option, please order in conjunction with environmental chamber
  • Choose based on size of chamber: small (for 311 L / 11 cu. ft. models) or large (for 821 L / 29 cu.ft. models)
  • Optional inner glass door enables visual check of samples with no major disturbance of temperature / humidity levels
TRUSTED_SUSTAINABILITY
Specifications
Description Solid door for large chamber, replaces standard outer glass door, factory installed
For Use With (Equipment) order with chamber models 3920, 3940, 3948, 3960, 3962
Lighting LED
Type Solid Door Option
Door Style Solid door for large chamber
For Use With (Application) light sensitive applications
Includes Factory installed solid door