Learn More
Thermo Scientific™ Solid Door Option with Inner Glass Door for Small Chamber
Product Code. 16401135 Shop All Thermo Scientific Products
Change view
Click to view available options
Door Style:
Solid with Inner Glass
Unit Size:
Each
Product Code. Door Style Unit Size
16401135 Solid with Inner Glass Each
1 options
Product Code. 16401135 Supplier Thermo Scientific™ Supplier No. 1900668
  • ONLINE EXCLUSIVE PRICE
    2550.00 €
    2460.00 € / Pack of 1
    Save 90.00 €
    3% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
Protection for your light sensitive samples
Solid outer door for Thermo Scientific™ chamber - replaces standard outer glass door to protect samples against light. Recommended for light sensitive applications. Factory-installed option, please order in conjunction with environmental chamber. F
  • Replaces standard outer glass door to protect samples against light
  • Recommended for light sensitive applications
  • Factory-installed option, please order in conjunction with environmental chamber
  • Choose based on size of chamber: small (for 311 L / 11 cu. ft. models) or large (for 821 L / 29 cu.ft. models)
  • Optional inner glass door enables visual check of samples with no major disturbance of temperature / humidity levels
TRUSTED_SUSTAINABILITY
Specifications
Description Solid door with inner glass door for small chamber, replaces standard outer glass door, factory installed
For Use With (Equipment) Chamber models 3911, 3907
Lighting LED
Type Solid Door Option
Door Style Solid with Inner Glass
For Use With (Application) Incubation, light sensitive applications
Includes Factory installed solid door with inner glass door