Learn More
AMPri™ SolidSafety ChemN Special Nitrile Gloves
Product Code. 16637405
Change view
Click to view available options
Size:
Large
X-Large
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16637405 Large Pack of 100
16667405 X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16637405 Supplier AMPri™ Supplier No. 81303L
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
SolidSafety ChemN Special Nitrile Protective Gloves are silicone-free and made of a special polymer that provides good chemical and physical resistance, as well as proven resistance to microorganisms.
  • Category III
  • Medical device Class I
  • Food grade
  • Chemical and physical resistance
  • Proven resistance to microorganisms
  • Silicone-free
  • Soft, comfortable fit, wearing comfort
  • Vera good grip, even if used in damp or wet environments
  • Non-sterile
  • Powder-free
  • Color: green
Specifications
Certifications/Compliance (Europe) EN 420, EN 455, EN 374/Type B (JKPT), Food grade according to Regulation (EU) 1935/2004, PPE Category III, Medical device Class I according to Regulation (EU) 2016/745
AQL 0.65
Color Green
Material Nitrile
Powdered Powder Free
Size Large
Length (Metric) 240 mm