missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Solus™ Anti-Fog / Anti-Scratch Coating (K&N)
Click to view available options
Lens Tint:
Clear
Gray
Includes:
Foam Gasket
n.a.
Unit Size:
Pack of 20
Description
- Safety glasses with a Corded Control System (CCS) on the temples for attaching corded ear plugs
- Scotchgard™ Anti-Fog/Anti-Scratch Coating resists fogging longer than traditional anti-fog coatings, even after up to 25 washings with water
- Anti-scratch coating provides increased durability
- 3M™ Scotchgard™ Anti-Fog/Anti-Scratch Coating provides superior anti-fog and anti-scratch properties, complying with K&N marking requirements of EN166
- Polycarbonate lens absorbs 99.9% of UVA and UVB rays
- Lens marking: 2C-1.2 3M 1 FT K N
- Frame marking: 3M EN 166 FT CE
- 3M internal testing per EN168 test method when compared with traditional anti-fog coatings
- Removable foam-lined gasket helps limit eye exposure to dust and debris while providing added comfort and cushioning
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | Clear |
| Frame Color | Green |
| Includes | Foam Gasket |
| Model | SCCS01SGAF-GRN-F-EU |
| Quantity | 20 /Cs. |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |
| Frame Type | Frameless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction