missing translation for 'onlineSavingsMsg'
Learn More

Solution D

Product Code. 11544936
Click to view available options
Quantity:
1 mL
25 mL
Unit Size:
1mL
25mL
This item is not returnable. View return policy

Product Code. 11544936

Brand: Invitrogen K1157

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solution D

Solution D is an anion transport inhibitor, for cells such as CHO-K1, that can be used in conjunction with CCF2-AM and CCF4-AM-based studies to prevent active cellular export of the FRET-based substrates after loading.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Pharma & Biopharma; Protein Assays and Analysis; Cellular Imaging
Product Line GeneBLAzer
Quantity 25 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.