missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Solution dNTPs
Shop All Cytiva Products
Click to view available options
Product Type:
dATP
dCTP
dGTP
dTTP
dUTP
Quantity:
25 μm
100 μm
500 μm
Unit Size:
Each
Description
- Free from DNase, RNase, and nicking enzyme activity
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
Specifications
| Format | Liquid |
| pH | 7 |
| Product Type | dATP |
| Concentration | 100 mM |
| Purity | >99% |
| Quantity | 100 μm |
| Color | Colorless |
| For Use With (Application) | Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction