missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Solution dNTPs: PCR Nucleotide Mix

Product Code. 10764547 Shop All Cytiva Products
Change view
Click to view available options
Concentration:
2 mM
10 mM
25 mM
Quantity:
500 μL
500 μm
1 mL
Unit Size:
1 set
500µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration Quantity Unit Size
10764547 10 mM 500 μL 500µL
10754547 2 mM 1 mL 1 set
10258184 25 mM 500 μm 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10764547 Supplier Cytiva Supplier No. 28406564
  • ONLINE EXCLUSIVE PRICE
    118.83 €
    105.43 € / 500µL
    Save 13.40 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Free from DNase, RNase, and nicking enzyme activity

  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

Order Info

Custom formulation, testing, packaging, and labeling are available

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Product Type PCR Nucleotide Mix dNTP Mix (10mM each A,C,G,T)
Concentration 10 mM
Purity >99%
Quantity 500 μL
Color Colorless
For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.