missing translation for 'onlineSavingsMsg'
Learn More

Solvent blue 35, MP Biomedicals™

Product Code. 11410981 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410981 100 g 100g
11400981 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410981 Supplier MP Biomedicals Supplier No. 0215663580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17354-14-2
Molecular Formula C22H26N2O2
Molecular Weight (g/mol) 350.462
MDL Number MFCD00011714
InChI Key OCQDPIXQTSYZJL-UHFFFAOYSA-N
Synonym C.I. 61554, Sudan Blue II
PubChem CID 3766139
IUPAC Name 1,4-bis(butylamino)anthracene-9,10-dione
SMILES CCCCNC1=C2C(=C(C=C1)NCCCC)C(=O)C3=CC=CC=C3C2=O

Specifications

Formula Weight 350.5
Density 1.23g/mL
Quantity 100 g
Physical Form Powder
Chemical Name or Material Solvent Blue 35, Dye content: Approx. 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.