missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Solvex™ 37-695 Series Green Nitrile Immersion Gloves

Product Code. 12610872
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12610872

Brand: Ansell™ 37695080

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Designed to deliver optimal results in wet or dry work environments where chemical resistance is crucial.

  • Industrial gloves
  • High-comfort, chemical resistant glove for range of applications
  • Fully reusable, with unequaled abrasion resistance
  • Provides comfort for the wearer
  • Food Approved
  • Cotton flocking, reducing sweat for the wearer
  • High performance nitrile compound provides chemical resistance and strength
  • Offers good snag, puncture and abrasion protection
  • Suitable for heavy-duty cleaning applications
  • High levels of flexibility, comfort and dexterity

Applications:

  • All food processing applications that have food contact
  • Delivery of parts to production line
  • Filling, blending and charging of raw materials
  • Forging operations
  • General pumping work
  • Handling incoming goods
  • Handling outgoing goods: cans, vessels, bulkpack and cartons
  • Loading and unloading process equipment
  • Opening and draining pumps valves or lines
  • Opening furnaces draining pumps valves or lines and crackers BTX process
  • Painting
  • Supervision of running operation
  • Testing
  • Transferring liquids and solids between vessels and tanks and process equipment
  • Welding
TRUSTED_SUSTAINABILITY

Specifications

Back Material Nitrile
Cuff Material Nitrile
Applications Industrial
Color Green
Latex Free Yes
Material Nitrile
Product Type Immersion Gloves
Size 8
Thickness (Metric) 0.42 mm
Glove Cuff Color Green
Glove Cuff Style Gauntlet
Liner Material Flocked
Quantity 12 Pairs
Resistant To Chemical resistance
Series 37-695
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.