missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sonicated DNAs for Hybridization Studies

Product Code. 10723217 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Sonicated Calf Thymus DNA
Sonicated Salmon Sperm DNA
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
10723217 Sonicated Calf Thymus DNA Each
10733217 Sonicated Salmon Sperm DNA Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723217 Supplier Cytiva Supplier No. 27456301
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for blocking non-specific binding sites in hybridization experiments to reduce background. Sonicated DNAs for Hybridization Studies are purified by phenol extraction and freeze dried.

  • Fragmented DNA with a size range from 200-6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease
  • DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 3000 bp
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Product Type Sonicated Calf Thymus DNA
Content And Storage -20°C
Quantity 100/Pk.
For Use With (Application) Hybridisation experiments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.