missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sorbic acid

Product Code. 18798496
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18798496 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18798496 Supplier Tocris Bioscience™ Supplier No. 7119/100
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Activator of hFFA2-DREADDs

Sorbic acid is an activator of hFFA2-DREADDs. Inhibits forskolin-induced cAMP production (IC50 ∼ 1 μM) and promotes accumulation of inositol monophosphates (IC50 >1 μM) in cells expressing hFFA2-DREADD. Promotes the release of GLP-1 from colonic crypts derived from hFFA2-DREADD-expressing mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 E,4 E)-2,4-Hexenoic acid
CAS 110-44-1
Quantity 100 mg
Target DREADD Activators
Molecular Formula C6H8O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.