missing translation for 'onlineSavingsMsg'
Learn More

Sorbic Acid, Extra Pure, SLR, Fisher Chemical

Product Code. 10102260 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10102260 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10102260 Supplier Fisher Chemical Supplier No. S/7390/48
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 110-44-1
Melting Point 132°C
Color White
pH 3.3
Packaging Plastic powder jar
Molecular Formula C6H8O2
MDL Number MFCD00002703
Quantity 100 g
Synonym sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
InChI Key WSWCOQWTEOXDQX-MQQKCMAXSA-N
SMILES C\C=C\C=C\C(O)=O
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
Molecular Weight (g/mol) 112.13
PubChem CID 643460
ChEBI CHEBI:38358
Formula Weight 112.13g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Hexadienoic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.