missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbic acid, free acid, ≥99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Specifications
Specifications
| Color | Yellow |
| Density | 1.2g/cm3 at 20°C |
| Quantity | 100 g |
| Formula Weight | 112.13 |
| Percent Purity | ≥99% |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sorbic acid, free acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction