missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan monolaurate

Product Code. 11348615
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348615 1000 g 1kg
11338615 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348615 Supplier Thermo Scientific Alfa Aesar Supplier No. L12099.0B
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1338-39-2 | C18H34O6 | 346.46 g/mol

Sorbitan monolaurate is a detergent and monoester of lauric acid and hexitol anhydrides derived from sorbitol. It is used in a study to assess enhanced solubility and oral bioavailability of itraconazole. It is also used in a study to investigate the oxidation of unsaturated lipids in oil-in-water emulsions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan monolaurate is a detergent and monoester of lauric acid and hexitol anhydrides derived from sorbitol. It is used in a study to assess enhanced solubility and oral bioavailability of itraconazole. It is also used in a study to investigate the oxidation of unsaturated lipids in oil-in-water emulsions.

Solubility
Soluble in 2-ethoxyethanol, ethanol and methanol. Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1338-39-2
Molecular Formula C18H34O6
Molecular Weight (g/mol) 346.46
MDL Number MFCD00005365
InChI Key LWZFANDGMFTDAV-IOVMHBDKSA-N
Synonym Sorbitan monododecanoate; Span™ 20
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Density 1.032
Quantity 1000 g
Flash Point >110°C (230°F)
Refractive Index 1.474
Merck Index 14,8724
Solubility Information Soluble in 2-ethoxyethanol,ethanol and methanol. Not miscible or difficult to mix in water.
Formula Weight 346.47
Chemical Name or Material Sorbitan monolaurate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.