missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, Extra Pure, SLR, meets analytical specification of Ph.Eur., USP, Fisher Chemical

Product Code. 10459760 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10459760 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10459760 Supplier Fisher Chemical Supplier No. S/7400/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 50-70-4
Melting Point 95°C
Color White
pH 6 to 7
Packaging Plastic powder jar
Molecular Formula C6H14O6
MDL Number 4708
Quantity 500 g
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
SMILES C(C(C(C(C(CO)O)O)O)O)O
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.172
PubChem CID 5780
ChEBI CHEBI:17924
Formula Weight 182.17g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.