missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, Fisher BioReagents

Product Code. 10396733
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10396733 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10396733 Supplier Fisher BioReagents Supplier No. BP439500
 Restricted Item
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sorbitol is used in the preparation of isoelectric focusing gels.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-70-4
Melting Point 75°C
Color White
Density 1.489g/cm³
pH 6 to 7
Boiling Point 295°C
Assay Percent Range 91.0 to 100.5 %
Packaging Poly Bottle
Molecular Formula C6H14O6
Quantity 500 g
Merck Index 15, 8851
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
SMILES C(C(C(C(C(CO)O)O)O)O)O
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.172
PubChem CID 5780
ChEBI CHEBI:17924
Formula Weight 182.17
Percent Purity 91.0 to 100.5%
Physical Form Solid
Chemical Name or Material Sorbitol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.