missing translation for 'onlineSavingsMsg'
Learn More

3M™ Sound Examiner 400 Series Sound Level Meter SE 402-IS

Product Code. 15740038
Change view
Click to view available options
Battery Life:
18 hours
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Battery Life unitSize
15740038 18 hours Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15740038 Supplier 3M™ Supplier No. 7100015833

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3M™ Sound Examiner 400 Series Sound Level Meter SE 402-IS is an intrinsically safe meter engineered to accurately measure noise levels in highly variable environments.

  • Intrinsically Safe
  • SPL, LMAX, LMIN, Peak, LEQ/LAVG, Elapsed Time
  • Variable, user-selectable data logging intervals
  • Rechargeable lithium battery
  • Store numerous noise surveys in memory for download
  • Removable Type/Class 1 microphone
  • Class 2
  • Multilingual user interface
TRUSTED_SUSTAINABILITY

Specifications

Battery Life 18 hours
Dimensions 28.4 x 7.1 x 3.3 cm
Product Type Sound Level Meter
For Use With (Application) Area Monitoring (Sound Level Meter)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.