missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Class/Type 1 unit with 1/1 and 1/3 Octave RTA and standard accessories
- Quick keypad calibration
- Removable SD memory card(2) virtual broadband SLMs
- Complete session, study and time history data logging
- 4 x AA Akaline cell batteries
- Optional Nickel Metal Hydride (NiMH Cells) batteries available
Specifications
Specifications
| Battery Life | >10 hours |
| Certifications/Compliance | IEC 61326-1 (2005), IEC 61672-1 (2002), ANSI S1.4 (R2006), ANSI S1.43 (R2007), IEC 61260 (2001), ANSI S1.11 (R2009), CE, WEEE, RoHS |
| Data Logging | Yes |
| Dimensions | 28.2 x 7.9 x 4.1 cm |
| Product Type | Sound Level Meter |
| For Use With (Application) | Area Monitoring (Sound Level Meter) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Ser du en mulighed for forbedring?