missing translation for 'onlineSavingsMsg'
Learn More

3M™ SoundPro™ SE/DL Series Sound Level Meter SP-DL-2-1/3

Product Code. 15790028
Change view
Click to view available options
Battery Life:
>10 hours
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Battery Life unitSize
15790028 >10 hours Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15790028 Supplier 3M™ Supplier No. 7100010781

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3M™ SoundPro™ SE/DL Series Sound Level Meter SP-DL-2-1/3 provides Class 2 sound level monitoring and comprehensive data analysis.

  • Class/Type 2 unit with 1/1 and 1/3 Octave RTA and standard accessories
  • Quick keypad calibration
  • Removable SD memory card(2) virtual broadband SLMs
  • Complete session, study and time history data logging
  • 4 x AA Akaline cell batteries
  • Optional Nickel Metal Hydride (NiMH Cells) batteries available
TRUSTED_SUSTAINABILITY

Specifications

Battery Life >10 hours
Certifications/Compliance IEC 61326-1 (2005), IEC 61672-1 (2002), ANSI S1.4 (R2006), ANSI S1.43 (R2007), IEC 61260 (2001), ANSI S1.11 (R2009), CE, WEEE, RoHS
Dimensions 28.2 x 7.9 x 4.1 cm
Product Type Sound Level Meter
For Use With (Application) Area Monitoring (Sound Level Meter)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.