missing translation for 'onlineSavingsMsg'
Learn More

Soy PC, TRC

Product Code. 30417482
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417482 1 g -
30409691 10 g -
30414334 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417482 Supplier Toronto Research Chemicals Supplier No. TRCS6778051G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97281-47-5

Specifications

Molecular Formula C42 H82 N O7 P
Quantity 1 g
Synonym Phosphatidylcholine from soy
InChI Formula InChI=1S/C43H82NO7P/c1-7-9-11-13-15-17-19-21-22-24-26-28-30-32-34-36-43(46)51-41(40-50-52(6,47)49-38-37-44(3,4)5)39-48-42(45)35-33-31-29-27-25-23-20-18-16-14-12-10-8-2/h15,17,21-22,41H,6-14,16,18-20,23-40H2,1-5H3/b17-15-,22-21-
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=C)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC
IUPAC Name 2-[[3-hexadecanoyloxy-2-[(9Z,12Z)-octadeca-9,12-dienoyl]oxy-propoxy]-methylene-oxido-$l^{5}-phosphanyl]oxyethyl-trimethyl-ammonium
Molecular Weight (g/mol) 756.09
Formula Weight 755.5829
Chemical Name or Material L-alpha-Phosphatidylcholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.