missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Soybean Oil, TRC
Specifications
Specifications
| Chemical Name or Material | Soybean Oil |
| CAS | 8001-22-7 |
| Quantity | 50 mL |
| Synonym | Soy Bean Oil, Soy-bean oil','AMCO','Best One Soya, Bionatrol','CAP 18, CT 7000, Degummed soybean oil, Destiny HC, HY 3050, Imperial Oil, K 1000, Kofa oil, Lipovenos, Nisshin Soybean White Oil, Pro Tech Natur'l Spray Oil, RBD, S-RBD, Shogun C, Soya fat, Vegetoil, Wesson |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?