missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SP 141

Description
SP 141 is a high affinity MDM2 inhibitor (Ki = 28 nM); directly binds MDM2. SP 141 inhibits MDM2 expression and induces MDM2 autoubiquitination and proteasomal degradation. SP 141 attenuates growth of breast cancer xenograft tumors. SP 141 impairs liposarcoma growth in vitro and in human xenograft models. SP 141 is cell permeable.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SP 141 |
| Purity | >98% |
| Molecular Weight (g/mol) | 324.38 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?